accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 551942-41-7

IUPAC Name: 4-[(diaminomethylidene)amino]-3-acetamido-2-(1,2,3-trihydroxypropyl)-3,4-dihydro-2H-pyran-6-carboxylic acid hydrate
Synonyms: 4-[(diaminomethylidene)amino]-5-acetamido-6-(1,2,3-trihydroxypropyl)-5,6-dihydro-4H-pyran-2-carboxylic acid hydrate
Molecular Weight (g/mol): 350.33
Molecular Formula: C12H22N4O8
InChi Key: OELRRAURPSTFEX-UHFFFAOYNA-N
SMILES: O.CC(=O)NC1C(C=C(OC1C(O)C(O)CO)C(O)=O)N=C(N)N
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results