Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zanamivir Hydrate 98.0+%, TCI America™
SDP

Catalog No. Z0023100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0023100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z0023100MG Supplier TCI America Supplier No. Z0023100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 551942-41-7
Molecular Formula C12H22N4O8
Molecular Weight (g/mol) 350.33
MDL Number MFCD00866966
InChI Key OELRRAURPSTFEX-UHFFFAOYNA-N
PubChem CID 636424
IUPAC Name 4-[(diaminomethylidene)amino]-3-acetamido-2-(1,2,3-trihydroxypropyl)-3,4-dihydro-2H-pyran-6-carboxylic acid hydrate
SMILES O.CC(=O)NC1C(C=C(OC1C(O)C(O)CO)C(O)=O)N=C(N)N

Specifications

Physical Form Powder/Crystals
Color White
Quantity 100 mg
Formula Weight 332.31
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Zanamivir Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.