accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOHNOO

CAS RN 5521-38-0

IUPAC Name: 2-[4-(4-nitrophenyl)piperazin-1-yl]ethan-1-ol
Synonyms: 2-[4-(4-nitrophenyl)piperazin-1-yl]ethanol
Molecular Weight (g/mol): 251.29
Molecular Formula: C12H17N3O3
InChi Key: WVNPWSRYQDRSIS-UHFFFAOYSA-N
SMILES: OCCN1CCN(CC1)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results