accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55276-43-2

IUPAC Name: 4-methanesulfonylpiperazin-1-ium
Synonyms: 4-methanesulfonylpiperazin-1-ium
Molecular Weight (g/mol): 165.23
Molecular Formula: C5H13N2O2S
InChi Key: ZZAKLGGGMWORRT-UHFFFAOYSA-O
SMILES: CS(=O)(=O)N1CC[NH2+]CC1
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)