accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNH2

CAS RN 55279-30-6

IUPAC Name: methyl 3-aminopyridine-4-carboxylate
Synonyms: methyl 3-aminopyridine-4-carboxylate
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: XLQIGLBALJNHKR-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=NC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)