Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Aminoisonicotinate 98.0+%, TCI America™
SDP

Catalog No. M26531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26531G Supplier TCI America Supplier No. M26531G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used for chemical research.

Chemical Identifiers

CAS 55279-30-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD02082535
InChI Key XLQIGLBALJNHKR-UHFFFAOYSA-N
Synonym methyl 3-aminoisonicotinate, 3-amino-isonicotinic acid methyl ester, 3-aminopyridine-4-carboxylic acid methyl ester, methyl3-aminoisonicotinate, 3-aminoisonicotinic acid methyl ester, 4-pyridinecarboxylic acid, 3-amino-, methyl ester, 3-amino-4-pyridinecarboxylic acid methyl ester, pubchem9760, acmc-1avm8, methyl 3-amino isonicotinate
PubChem CID 1488824
IUPAC Name methyl 3-aminopyridine-4-carboxylate
SMILES COC(=O)C1=C(C=NC=C1)N

Specifications

Melting Point 83°C
Color Yellow
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Aminoisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.