accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOSOOHNH2

CAS RN 55288-30-7

IUPAC Name: 2-amino-3-[(4-nitrophenyl)sulfanyl]propanoic acid
Synonyms: 2-amino-3-[(4-nitrophenyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 242.25
Molecular Formula: C9H10N2O4S
InChi Key: WXMIOWSCOYJQOT-UHFFFAOYNA-N
SMILES: NC(CSC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results