Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-(4-Nitrophenyl)-L-cysteine 98.0+%, TCI America™
SDP

Catalog No. N0612100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0612100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0612100MG Supplier TCI America Supplier No. N0612100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unnatural amino acid derivative for use as a laboratory reagent.

Chemical Identifiers

CAS 55288-30-7
Molecular Formula C9H10N2O4S
Molecular Weight (g/mol) 242.25
MDL Number MFCD00671568
InChI Key WXMIOWSCOYJQOT-UHFFFAOYNA-N
PubChem CID 15298820
IUPAC Name 2-amino-3-[(4-nitrophenyl)sulfanyl]propanoic acid
SMILES NC(CSC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Color Yellow
Packaging Glass Bottle
Quantity 100 mg
Formula Weight 242.25
Percent Purity ≥98.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material S-(4-Nitrophenyl)-L-cysteine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.