accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 553-17-3

IUPAC Name: bis(2-methoxyphenyl) carbonate
Synonyms: guaiacol carbonate
Molecular Weight (g/mol): 274.27
Molecular Formula: C15H14O5
InChi Key: ORUJFMPWKPVXLZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC(=O)OC1=CC=CC=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results