Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiacol Carbonate 98.0+%, TCI America™
SDP

Catalog No. C0039500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0039500G 500 g
C003925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0039500G Supplier TCI America Supplier No. C0039500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 553-17-3
Molecular Formula C15H14O5
Molecular Weight (g/mol) 274.272
MDL Number MFCD00025723
InChI Key ORUJFMPWKPVXLZ-UHFFFAOYSA-N
Synonym guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29
PubChem CID 11104
IUPAC Name bis(2-methoxyphenyl) carbonate
SMILES COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC

Specifications

Melting Point 88°C
Color White
Quantity 500 g
Formula Weight 274.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Guaiacol Carbonate
EINECSNumber (9)-0330
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.