accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55302-96-0

IUPAC Name: 5-[(2-hydroxyethyl)amino]-2-methylphenol
Synonyms: 5-[(2-hydroxyethyl)amino]-2-methylphenol
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: YGRFRBUGAPOJDU-UHFFFAOYSA-N
SMILES: CC1=CC=C(NCCO)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results