Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Hydroxyethyl)amino-o-cresol 98.0+%, TCI America™
SDP

Catalog No. H138125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H138125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H138125G Supplier TCI America Supplier No. H138125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55302-96-0
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD01707346
InChI Key YGRFRBUGAPOJDU-UHFFFAOYSA-N
Synonym 5-(2-Hydroxyethyl)amino-2-methylphenol
PubChem CID 134396
IUPAC Name 5-(2-hydroxyethylamino)-2-methylphenol
SMILES CC1=C(C=C(C=C1)NCCO)O

Specifications

Melting Point 93°C
Quantity 25 g
Formula Weight 167.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Hydroxyethyl)amino-o-cresol
RTECSNumber SL5078600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.