accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2N

CAS RN 55338-73-3

IUPAC Name: 5-aminopyridine-2-carbonitrile
Synonyms: 5-aminopyridine-2-carbonitrile
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: IFOXWHQFTSCNQB-UHFFFAOYSA-N
SMILES: NC1=CN=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results