CAS RN 5534-09-8
IUPAC Name:
2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-chloro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
Synonyms:
beconase Spir beclovent beclomethasone dipropionate andion beclate vancenase vanceril becotide beclometasone dipropionate
Molecular Weight (g/mol):
521.05
Molecular Formula:
C28H37ClO7
InChi Key:
KUVIULQEHSCUHY-XYWKZLDCSA-N
SMILES:
CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
1
–
1
of
1
results
1
Medisca Beclomethasone Dipropionate, USP (Micronized)
Beclomethasone Dipropionate USP (Micronized) is a white to cream white, odorless powder. Micronized with a particle size distribution of 90% ≤ 5 μm.