Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Beclomethasone Dipropionate, USP (Micronized)
SDP

Catalog No. 14185418 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-418 1 g
14-185-409 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-185-418 Supplier Medisca Supplier No. 036406
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beclomethasone Dipropionate USP (Micronized) is a white to cream white, odorless powder. Micronized with a particle size distribution of 90% ≤ 5 μm.

Beclomethasone Dipropionate USP (Micronized) is a white to cream white, odorless powder. Micronized with a particle size distribution of 90% ≤ 5 μm.

Alternative name(s): Beclometasone Dipropionate

Specifications

Chemical Name or Material Beclomethasone Dipropionate, USP (Micronized)
Packaging Bottle, 15mL, Amber Glass Wide Mouth Packer, 28-400
CAS 5534-09-8
Type Drug Standard
Molecular Weight (g/mol) 521.04
Quantity 1 g
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.