accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5537-71-3

IUPAC Name: 3-(1-cyanoethyl)benzoic acid
Synonyms: 3-(1-cyanoethyl)benzoic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: IRYIYPWRXROPSX-UHFFFAOYNA-N
SMILES: CC(C#N)C1=CC=CC(=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results