accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClO

CAS RN 5538-51-2

IUPAC Name: 2-(carbonochloridoyl)phenyl acetate
Synonyms: 2-(carbonochloridoyl)phenyl acetate
Molecular Weight (g/mol): 198.6
Molecular Formula: C9H7ClO3
InChi Key: DSGKWFGEUBCEIE-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C(Cl)=O
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)