accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55387-85-4

IUPAC Name: 2-(2,2-diethoxyethyl)propane-1,3-diol
Synonyms: 2-(2,2-diethoxyethyl)propane-1,3-diol
Molecular Weight (g/mol): 192.26
Molecular Formula: C9H20O4
InChi Key: HKZYWLNVGYRMDG-UHFFFAOYSA-N
SMILES: CCOC(CC(CO)CO)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results