accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 554-91-6

IUPAC Name: (2R,3R,4S,5S,6R)-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-2,3,4,5-tetrol
Synonyms: gentiobiose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: DLRVVLDZNNYCBX-LIZSDCNHSA-N
SMILES: OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)