Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentiobiose 97%, Thermo Scientific™

Catalog No. AC225912500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225912500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225912500 Supplier Thermo Scientific Chemicals Supplier No. 225912500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-91-6
Molecular Formula C12H22O11
Synonym 6-O-β-D-Glucopyranosyl-D-glucopyranose

Specifications

Color White
Loss on Drying 1% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Quantity 250 mg
Beilstein 31, 397
Merck Index 15, 4431
Specific Rotation After 24 hrs.: +9° to +11° (20°C, 589nm) (c=2, water)
Formula Weight 342.29
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material Gentiobiose, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.