CAS RN 55465-40-2
IUPAC Name:
aluminium(3+) sodium bis(oxosilanebis(olate))
Synonyms:
sodium aluminosilicate sodium silicoaluminate
Molecular Weight (g/mol):
202.14
Molecular Formula:
AlNaO6Si2
InChi Key:
URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES:
[Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
1
–
1
of
1
results
1
Molecular Sieves, Grade 564 3 Angstroms) 8-12 Mesh), Macron Fine Chemicals™
CAS: 55465-40-2 Molecular Formula: AlNaO6Si2 Molecular Weight (g/mol): 202.135 InChI Key: URGAHOPLAPQHLN-UHFFFAOYSA-N PubChem CID: 19758701 IUPAC Name: aluminum;sodium;dioxido(oxo)silane SMILES: [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]