accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55489-58-2

IUPAC Name: 2-{2-[2-(benzyloxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: 2-{2-[2-(benzyloxy)ethoxy]ethoxy}ethanol
Molecular Weight (g/mol): 240.3
Molecular Formula: C13H20O4
InChi Key: KVPHZILZNXDCNH-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results