Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Monobenzyl Ether 97.0+%, TCI America™
SDP

Catalog No. T19965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19965G 5 g
T199625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19965G Supplier TCI America Supplier No. T19965G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55489-58-2
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.299
MDL Number MFCD02093556
InChI Key KVPHZILZNXDCNH-UHFFFAOYSA-N
Synonym 2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethanol
PubChem CID 2737322
IUPAC Name 2-[2-(2-phenylmethoxyethoxy)ethoxy]ethanol
SMILES C1=CC=C(C=C1)COCCOCCOCCO

Specifications

Color Yellow
Quantity 5 g
Formula Weight 240.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Monobenzyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.