accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 555-31-7

IUPAC Name: aluminium(3+) tris(propan-2-olate)
Synonyms: aluminium isopropoxide
Molecular Weight (g/mol): 204.25
Molecular Formula: C9H21AlO3
InChi Key: SMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES: [Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Molecular Weight (g/mol) 
  • (5)
  • (6)
  • (6)
Percent Purity 
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (6)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (6)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (2)
  • (10)
  • (5)
  • (6)
  • (6)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)