CAS RN 555-35-1
IUPAC Name:
aluminium(3+) trihexadecanoate
Synonyms:
aluminium(3+) tris(n-hexadecanoate)
Molecular Weight (g/mol):
793.25
Molecular Formula:
C48H93AlO6
InChi Key:
PZWDHVKNXVLHOV-UHFFFAOYSA-K
SMILES:
[Al+3].CCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCC([O-])=O
1
–
1
of
1
results
1
MP Biomedicals, Inc Aluminum Palmitate, MP Biomedicals
CAS: 555-35-1 Molecular Formula: C48H96AlO6 Molecular Weight (g/mol): 796.272 InChI Key: JJCSYJVFIRBCRI-UHFFFAOYSA-N Synonym: aluminum palmitate PubChem CID: 131881080 IUPAC Name: aluminum;hexadecanoic acid SMILES: CCCCCCCCCCCCCCCC(=O)O.CCCCCCCCCCCCCCCC(=O)O.CCCCCCCCCCCCCCCC(=O)O.[Al]