Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Palmitate, MP Biomedicals

Catalog No. ICN206474 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206474 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206474 Supplier MP Biomedicals, Inc Supplier No. 0520647480

Chemical Identifiers

CAS 555-35-1
Molecular Formula C48H96AlO6
Molecular Weight (g/mol) 796.272
InChI Key JJCSYJVFIRBCRI-UHFFFAOYSA-N
Synonym aluminum palmitate
PubChem CID 131881080
IUPAC Name aluminum;hexadecanoic acid
SMILES CCCCCCCCCCCCCCCC(=O)O.CCCCCCCCCCCCCCCC(=O)O.CCCCCCCCCCCCCCCC(=O)O.[Al]

Specifications

Quantity 100 g
Formula Weight 793.25
Chemical Name or Material Aluminum Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.