accessibility menu, dialog, popup

UserName

Viewing profile as user:
Si

CAS RN 5550-08-3

IUPAC Name: 8,8-diphenyl-8-silatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene
Synonyms: 8,8-diphenyl-8-silatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene
Molecular Weight (g/mol): 334.49
Molecular Formula: C24H18Si
InChi Key: RVHKZECCDOKCIB-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Si]1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results