Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Diphenyl-9H-9-silafluorene 98.0+%, TCI America™
SDP

Catalog No. D5034200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5034200MG 200 mg
D50341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5034200MG Supplier TCI America Supplier No. D5034200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5550-08-3
Molecular Formula C24H18Si
Molecular Weight (g/mol) 334.49
InChI Key RVHKZECCDOKCIB-UHFFFAOYSA-N
Synonym 5,5-Diphenyl-5H-dibenzo[b,d]silole
PubChem CID 424835
IUPAC Name 5,5-diphenylbenzo[b][1]benzosilole
SMILES C1=CC=C(C=C1)[Si]2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=CC=C5

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 200 mg
Formula Weight 334.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Diphenyl-9H-9-silafluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.