accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)NNHN(R)OP(R)(S)OOOOOH2NOHHONaNa

CAS RN 5550-12-9

IUPAC Name: disodium [(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Synonyms: guanylate
Molecular Weight (g/mol): 407.19
Molecular Formula: C10H12N5Na2O8P
InChi Key: PVBRXXAAPNGWGE-KHRSEZDTNA-L
SMILES: [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)