accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5552-44-3

IUPAC Name: 2,5-diethyl pyridine-2,5-dicarboxylate
Synonyms: 2,5-diethyl pyridine-2,5-dicarboxylate
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: AFWWHZBUQWEDLS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(N=C1)C(=O)OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results