Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2,5-Pyridinedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. P109410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P109410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P109410G Supplier TCI America Supplier No. P109410G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic reagent with a purity of >98.0% determine by GC.

Chemical Identifiers

CAS 5552-44-3
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00145190
InChI Key AFWWHZBUQWEDLS-UHFFFAOYSA-N
Synonym 2,5-Pyridinedicarboxylic Acid Diethyl Ester
PubChem CID 254728
IUPAC Name 2,5-diethyl pyridine-2,5-dicarboxylate
SMILES CCOC(=O)C1=CC=C(N=C1)C(=O)OCC

Specifications

Melting Point 46°C
Boiling Point 181°C
Quantity 10 g
Formula Weight 223.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 2,5-Pyridinedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.