accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSHNN(E)

CAS RN 5554-99-4

IUPAC Name: (2E)-2-(1H-1,3-benzodiazol-2-yl)-3-(thiophen-2-yl)prop-2-enenitrile
Synonyms: (2E)-2-(1H-1,3-benzodiazol-2-yl)-3-(thiophen-2-yl)prop-2-enenitrile
Molecular Weight (g/mol): 251.31
Molecular Formula: C14H9N3S
InChi Key: MDVSLHGZTWHJMW-CSKARUKUSA-N
SMILES: N#C\C(=C/C1=CC=CS1)C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results