accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55557-52-3

IUPAC Name: 3-chloropyrazine-2-carbonitrile
Synonyms: 3-chloropyrazine-2-carbonitrile
Molecular Weight (g/mol): 139.54
Molecular Formula: C5H2ClN3
InChi Key: SDLFAEGTVBPHBK-UHFFFAOYSA-N
SMILES: ClC1=NC=CN=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results