accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OH

CAS RN 556-82-1

IUPAC Name: 3-methylbut-2-en-1-ol
Synonyms: prenol dimethylallyl alcohol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: ASUAYTHWZCLXAN-UHFFFAOYSA-N
SMILES: CC(C)=CCO
Boiling Point 
  • (5)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (6)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (3)
  • (8)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (8)