accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOOOHOHOOHO

CAS RN 5560-59-8

IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid; ethylbis(3-phenylpropyl)amine
Synonyms: alverine
Molecular Weight (g/mol): 473.57
Molecular Formula: C26H35NO7
InChi Key: RYHCACJBKCOBTJ-UHFFFAOYSA-N
SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CCCC1=CC=CC=C1)CCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results