Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alverine, MP Biomedicals™

Catalog No. ICN15473225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15473225 25 g
ICN15473205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15473225 Supplier MP Biomedicals, Inc Supplier No. 0215473225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5560-59-8
Molecular Formula C26H35NO7
Molecular Weight (g/mol) 473.566
InChI Key RYHCACJBKCOBTJ-UHFFFAOYSA-N
Synonym alverine citrate, alverine citrate salt, nci 85x, unii-9jfb58yk1e, 9jfb58yk1e, n-ethyl-3,3'-diphenyldipropylamine citrate 1:1, dsstox_cid_25562, dsstox_rid_80959, dsstox_gsid_45562
PubChem CID 21718
ChEBI CHEBI:53785
IUPAC Name N-ethyl-3-phenyl-N-(3-phenylpropyl)propan-1-amine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CCCC1=CC=CC=C1)CCCC2=CC=CC=C2.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Quantity 25 g
Formula Weight 473.6
Physical Form Powder
Chemical Name or Material Alverine
EINECSNumber 226-929-3
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.