accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55628-54-1

IUPAC Name: (2R,3S,4S)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
Synonyms: (2R,3S,4S)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
Molecular Weight (g/mol): 416.52
Molecular Formula: C27H28O4
InChi Key: MXYLLYBWXIUMIT-VJGNERBWSA-N
SMILES: C(OCC1=CC=CC=C1)[C@H]1OC=C[C@H](OCC2=CC=CC=C2)[C@@H]1OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)