Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-2-Heptanol

Catalog No. 3403321G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-033-21G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 34-033-21G Supplier Sigma Aldrich Supplier No. 3403321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-2-Heptanol can be used as a starting material to synthesize:(S)-(+)-2-benzoyloxyheptane by treating with benzoyl anhydride and Et3N in the presence of DMAP (S)-2-(p-toluenesulfonyl)heptane by reacting with p-toluenesulfonyl chloride (+)-2-Heptyl acetate by treating with acetyl chloride.

Specifications

CAS 55628-54-1
Melting Point 57°C to 58°C
Boiling Point 62°C (20 mmHg, lit.)
Molecular Formula C27H28O4
Refractive Index n20/D 1.421
Linear Formula C27H28O4
MDL Number MFCD00061640
Quantity 5 g
Molecular Weight (g/mol) 416.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.