accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnOOCH2OOCH2

CAS RN 557-08-4

IUPAC Name: zinc(2+) bis(undec-10-enoate)
Synonyms: zinc(2+) bis(undec-10-enoate)
Molecular Weight (g/mol): 431.92
Molecular Formula: C22H38O4Zn
InChi Key: YMCOHQVWOBMDCZ-UHFFFAOYSA-L
SMILES: [Zn++].[O-]C(=O)CCCCCCCCC=C.[O-]C(=O)CCCCCCCCC=C
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)
Percent Purity 
  • (5)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (5)