Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Undecylenate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607892 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-892 25 kg Amber Glass Bottle
18-607-889 500 g Amber Glass Bottle
18-607-890 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-892 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ZI12525KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ZI125, 557-08-4, C22H38O4Zn

Spectrum™ Chemical Zinc Undecylenate, USP is used to treat skin fungal infections such as athlete's foot and jock itch. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-08-4
Molecular Formula C22H38O4Zn
Molecular Weight (g/mol) 431.92
InChI Key YMCOHQVWOBMDCZ-UHFFFAOYSA-L
IUPAC Name zinc(2+) bis(undec-10-enoate)
SMILES [Zn++].[O-]C(=O)CCCCCCCCC=C.[O-]C(=O)CCCCCCCCC=C

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 25 kg
Percent Purity 98 to 102%
Grade USP
Loss on Drying 0.0125
Packaging Amber Glass Bottle
Chemical Name or Material Zinc Undecylenate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.