accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 557-40-4

IUPAC Name: 3-(prop-2-en-1-yloxy)prop-1-ene
Synonyms: allyl ether
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: ATVJXMYDOSMEPO-UHFFFAOYSA-N
SMILES: C=CCOCC=C
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)