accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 55726-23-3

IUPAC Name: docosanoyl docosanoate
Synonyms: docosanoyl docosanoate
Molecular Weight (g/mol): 663.17
Molecular Formula: C44H86O3
InChi Key: IJVLPVWXJLKHID-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)