Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Behenic Anhydride 97.0+%, TCI America™
SDP

Catalog No. B15315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15315G 5 g
B15311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15315G Supplier TCI America Supplier No. B15315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55726-23-3
Molecular Formula C44H86O3
Molecular Weight (g/mol) 663.17
MDL Number MFCD00010452
InChI Key IJVLPVWXJLKHID-UHFFFAOYSA-N
Synonym Docosanoic Anhydride
PubChem CID 566696
IUPAC Name docosanoyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 5 g
Formula Weight 663.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Behenic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.