accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiClCl

CAS RN 5578-42-7

IUPAC Name: dichloro(cyclohexyl)methylsilane
Synonyms: dichloro(cyclohexyl)methylsilane
Molecular Weight (g/mol): 197.17
Molecular Formula: C7H14Cl2Si
InChi Key: YUYHCACQLHNZLS-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)C1CCCCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)