Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorocyclohexylmethylsilane 97.0+%, TCI America™
SDP

Catalog No. C13995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13995G 5 g
C139925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13995G Supplier TCI America Supplier No. C13995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5578-42-7
Molecular Formula C7H14Cl2Si
Molecular Weight (g/mol) 197.174
MDL Number MFCD00046333
InChI Key YUYHCACQLHNZLS-UHFFFAOYSA-N
Synonym methylcyclohexyldichlorosilane, silane, dichlorocyclohexylmethyl, dichloro cyclohexyl methyl silane, cyclohexylmethyldichlorosilane, cyclohexane, dichloromethylsilyl, methyl cyclohexyldichlorosilane, cyclohexyl methyl dichlorosilane, cyclohexyl-dichloro-methylsilane, cyclohexyl-methyl-dichlorosilane, dichloro cyclohexyl methylsilane
PubChem CID 79685
IUPAC Name dichloro-cyclohexyl-methylsilane
SMILES C[Si](C1CCCCC1)(Cl)Cl

Specifications

Color Colorless
Boiling Point 202°C
Quantity 5 g
UN Number 2987
Formula Weight 197.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorocyclohexylmethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.