accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55780-92-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-methylpentanoic acid; dicha
Molecular Weight (g/mol): 412.62
Molecular Formula: C23H44N2O4
InChi Key: NCLUKCZVJYNXBY-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CCC(C)C(NC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)