Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-D-allo-isoleucine dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog No. AAH66824MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66824MD 250 mg
AAH6682403 1 g
AAH6682406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66824MD Supplier Thermo Scientific Chemicals Supplier No. H66824MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55780-92-2
Molecular Formula C23H44N2O4
Molecular Weight (g/mol) 412.62
MDL Number MFCD28411344
InChI Key NCLUKCZVJYNXBY-UHFFFAOYNA-N
Synonym boc-d-allo-ile-oh.dcha
PubChem CID 122173517
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoic acid; N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1.CCC(C)C(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Quantity 250 mg
Formula Weight 412.62
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-D-allo-isoleucine dicyclohexylammonium salt
storageNote1 -30° to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.