accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOOOOOOOOH

CAS RN 5579-66-8

IUPAC Name: 3,6,9,12,15,18,21,24,27-nonaoxanonacosane-1,29-diol
Synonyms: decaethylene glycol polyethylene glycol 500 polyoxyethylene (10)
Molecular Weight (g/mol): 458.55
Molecular Formula: C20H42O11
InChi Key: DTPCFIHYWYONMD-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)