Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decaethylene Glycol 90.0+%, TCI America™
SDP

Catalog No. D29001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29001G Supplier TCI America Supplier No. D29001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5579-66-8
Molecular Formula C20H42O11
Molecular Weight (g/mol) 458.545
MDL Number MFCD00698693
InChI Key DTPCFIHYWYONMD-UHFFFAOYSA-N
PubChem CID 79689
ChEBI CHEBI:46550
IUPAC Name 2-[2-[2-[2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCOCCOCCOCCOCCOCCO)O

Specifications

Melting Point 28°C
Color White-Yellow
Boiling Point 280°C
Quantity 1 g
Formula Weight 458.55
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Decaethylene Glycol
EINECSNumber (2)-0441
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.