accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClH3CNHN

CAS RN 5579-84-0

IUPAC Name: dihydrogen methyl[2-(pyridin-2-yl)ethyl]amine dichloride
Synonyms: dihydrogen betahistine dichloride
Molecular Weight (g/mol): 209.11
Molecular Formula: C8H14Cl2N2
InChi Key: XVDFMHARQUBJRE-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CNCCC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (4)